Skip to Content
Merck
All Photos(1)

Key Documents

425443

Sigma-Aldrich

Benzamide-15N

98 atom % 15N

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$650.00

$650.00


Estimated to ship onApril 16, 2025


Request more information

Select a Size

Change View
1 G
$650.00

About This Item

Linear Formula:
C6H5CO15NH2
CAS Number:
Molecular Weight:
122.13
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$650.00


Estimated to ship onApril 16, 2025


Request more information

isotopic purity

98 atom % 15N

Quality Level

Assay

99% (CP)

form

solid

mp

125-128 °C (lit.)

mass shift

M+1

SMILES string

[15NH2]C(=O)c1ccccc1

InChI

1S/C7H7NO/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H2,8,9)/i8+1

InChI key

KXDAEFPNCMNJSK-VJJZLTLGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service