Skip to Content
Merck
All Photos(1)

Key Documents

488143

Sigma-Aldrich

1,2-Bis[bis(pentafluorophenyl)phosphino]ethane

Synonym(s):

1,2-Bis(dipentafluorophenylphosphino)ethane, 1,2-Ethanediylbis[bis(pentafluorophenyl)phosphine, Bis(dipentafluorophenylphosphine)ethane, Dfppe

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6F5)2PCH2-]2
CAS Number:
Molecular Weight:
758.23
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

192-195 °C (lit.)

functional group

phosphine

SMILES string

Fc1c(F)c(F)c(P(CCP(c2c(F)c(F)c(F)c(F)c2F)c3c(F)c(F)c(F)c(F)c3F)c4c(F)c(F)c(F)c(F)c4F)c(F)c1F

InChI

1S/C26H4F20P2/c27-3-7(31)15(39)23(16(40)8(3)32)47(24-17(41)9(33)4(28)10(34)18(24)42)1-2-48(25-19(43)11(35)5(29)12(36)20(25)44)26-21(45)13(37)6(30)14(38)22(26)46/h1-2H2

InChI key

IGLFIYOFKVGEBP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service