Skip to Content
Merck
All Photos(2)

Key Documents

109223

Sigma-Aldrich

1-Bromoadamantane

99%

Synonym(s):

1-Adamantyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15Br
CAS Number:
Molecular Weight:
215.13
Beilstein:
1098857
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

mp

116-118 °C (lit.)

functional group

bromo

SMILES string

BrC12C[C@H]3C[C@H](C[C@H](C3)C1)C2

InChI

1S/C10H15Br/c11-10-4-7-1-8(5-10)3-9(2-7)6-10/h7-9H,1-6H2/t7-,8+,9-,10-

InChI key

VQHPRVYDKRESCL-CHIWXEEVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromoadamantane forms inclusion complexes with α-, β-, and γ-cyclodextrins[1]. It causes the alkylation of Co(II) complexes of β-diketones[2].

Application

1-Bromoadamantane was used in the synthesis of a new organic–organic nanoscale composite thin-film dielectric[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P. M. Ivanov et al.
The Journal of organic chemistry, 61(20), 7012-7017 (1996-10-04)
The inclusion complexes between the most commonly used cyclodextrins (alpha-, beta-, and gamma-CD) and 1-bromoadamantane were prepared and studied experimentally by NMR methods and by molecular dynamics simulations (AMBER force field) with solvation. The NMR results suggest host/guest ratios of
Jitendra N Tiwari et al.
ChemSusChem, 3(9), 1051-1056 (2010-07-14)
A new organic-organic nanoscale composite thin-film (NCTF) dielectric has been synthesized by solution deposition of 1-bromoadamantane and triblock copolymer (Pluronic P123, BASF, EO20-PO70-EO20), in which the precursor solution has been achieved with organic additives. We have used a sol-gel process
Alkylation of ?-diketones through their co (II), co (III) and zn (II) complexes. 1-Bromoadamantane as alkylating agent.
Gonzalez A, et al.
Tetrahedron Letters, 26(31), 3735-3738 (1985)
Cheng Li et al.
Analytica chimica acta, 1066, 121-130 (2019-04-28)
A visual, rapid, and sensitive method for the detection of two algal metabolites, geosmin (GSM) and 2-methylisoborneol (2-MIB) using a competitive displacement technique based on molecular imprinted polymers (MIPs) and fluorescent tags was developed. In this method, fluorescent tags that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service