Skip to Content
Merck
All Photos(1)

Key Documents

487805

Sigma-Aldrich

Sodium acetate-18O2

95 atom % 18O

Synonym(s):

Acetic acid-18O2 sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$3,650.00

$3,650.00


Estimated to ship onMay 25, 2025


Request more information

Select a Size

Change View
1 G
$3,650.00

About This Item

Linear Formula:
CH3C18O2Na
CAS Number:
Molecular Weight:
86.03
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

$3,650.00


Estimated to ship onMay 25, 2025


Request more information

isotopic purity

95 atom % 18O

form

solid

mp

>300 °C (dec.) (lit.)

mass shift

M+4

SMILES string

[Na+].CC([18O-])=[18O]

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1/i3+2,4+2;

InChI key

VMHLLURERBWHNL-MZCPDTSISA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service