Skip to Content
Merck
All Photos(4)

Key Documents

482307

Sigma-Aldrich

Silver trifluoroacetate

≥99.99% trace metals basis

Synonym(s):

Trifluoroacetic acid silver salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$93.60
2.5 G
$213.00
10 G
$607.00

$93.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$93.60
2.5 G
$213.00
10 G
$607.00

About This Item

Linear Formula:
CF3COOAg
CAS Number:
Molecular Weight:
220.88
Beilstein:
3634022
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$93.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99.99% trace metals basis

form

crystalline powder
powder, crystals or chunks

reaction suitability

core: silver

mp

257-260 °C (dec.) (lit.)

SMILES string

FC(F)(F)C(=O)O[Ag]

InChI

1S/C2HF3O2.Ag/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1

InChI key

KZJPVUDYAMEDRM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Silver trifluoroacetate is a versatile reagent used in organic synthetic reactions, including acylation, alkylation, and cyclization reactions. It is also used to fabricate silver nanomaterials.[1]

Application

Silver trifluoroacetate can be used:
  • As a precursor to synthesize silver nanoparticles.[1]
  • As a starting material to fabricate conducting silver networks with high transmittance and low resistance via a polymer-assisted electrospinning technique.[2]
  • As a reagent to prepare polynuclear luminescent complexes.[3]
  • As a bifunctional reagent for oxidative carbonylative [4 + 1] annulation of aromatic acid.[4]

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct Synthesis of Narrowly Dispersed Silver Nanoparticles Using a Single-Source Precursor
Xue Zhang Lin, et al.
Langmuir, 19, 10081-10085 (2003)
Silver trifluoroacetate as a bifunctional reagent for palladium-catalyzed oxidative carbonylative [4+ 1] annulation of aromatic acids
Zhuang-Zhuang Li, et al
Organic Chemistry Frontiers : An International Journal of Organic Chemistry / Royal Society of Chemistry, 10, 2243-2250 (2023)
Perhalophenyl(tetrahydrothiophene)gold(I) Complexes as Lewis Bases in Acid?Base Reactions with Silver Trifluoroacetate
Eduardo J. Fernandez, et al.},
Organometallics, 26, 5931-5939 (2007)
Conducting Silver Networks Based on Electrospun Poly(Methyl Methacrylate) and Silver Trifluoroacetate
Hung-Tao Chen, et al.
ACS Applied Materials & Interfaces, 7, 9479-9485 (2015)

Articles

Copper metal deposition processes are an essential tool for depositing interconnects used in microelectronic applications, giving group 11 (coinage metals: Copper, Silver, and Gold) an important place in atomic layer deposition (ALD) process development.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service