Skip to Content
Merck
All Photos(1)

Key Documents

486612

Sigma-Aldrich

DL-Valine-d8

98 atom % D

Synonym(s):

DL-Valine-2,3,4,4,4,5,5,5-d8

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$2,360.00

$2,360.00


Estimated to ship onMay 24, 2025


Request more information

Select a Size

Change View
1 G
$2,360.00

About This Item

Linear Formula:
(CD3)2CDCD(NH2)CO2H
CAS Number:
Molecular Weight:
125.20
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

$2,360.00


Estimated to ship onMay 24, 2025


Request more information

isotopic purity

98 atom % D

Assay

99% (CP)

form

solid

mp

295 °C (subl.) (lit.)

mass shift

M+8

SMILES string

[2H]C([2H])([2H])C([2H])(C([2H])([2H])[2H])C([2H])(N)C(O)=O

InChI

1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/i1D3,2D3,3D,4D

InChI key

KZSNJWFQEVHDMF-PIODKIDGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Gray et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1106-1107, 50-57 (2019-01-15)
A UPLC-MS/MS assay, employing a reversed-phase separation, has been applied to the analysis of a number of common amino acids and biogenic amines in rat urine. Analytes were derivatised, using 6‑aminoquinolyl‑N‑hydroxysuccinimidyl carbamate (AccQTag Ultra™). Derivatisation with this reagent, by increasing
Zhi Zhou et al.
Analytica chimica acta, 1109, 44-52 (2020-04-08)
The development of quantitative metabolomics approaches for future standardized and translational applications has become increasingly important. Data-independent targeted quantitative metabolomics (DITQM) is a newly proposed method providing ion pair information on 1324 metabolites. However, the quantification of more than 1000

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service