Skip to Content
Merck
All Photos(1)

Documents

486515

Sigma-Aldrich

Deuterium bromide

99 atom % D

Synonym(s):

Hydrogen-d bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DBr
CAS Number:
Molecular Weight:
81.92
EC Number:
MDL number:
UNSPSC Code:
12352003
PubChem Substance ID:
NACRES:
NA.12

vapor density

2.71 (20 °C, vs air)

Quality Level

vapor pressure

334.7 psi ( 21 °C)

isotopic purity

99 atom % D

bp

−67 °C (lit.)

mp

−87 °C (lit.)

mass shift

M+1

SMILES string

Br[2H]

InChI

1S/BrH/h1H/i/hD

InChI key

CPELXLSAUQHCOX-DYCDLGHISA-N

Related Categories

Packaging

5 L quantity is packaged in a 450 mL carbon steel lecture bottle with stainless steel CGA 110/180 valve. Nominal gas pressure at 21ºC is 145 psig for 5 L. These pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
Shipping of this product by air freight is forbidden.
Outside of the U.S., ocean freight shipment is required and will result in substantial additional expense.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Monel control valve Z146978 or Monel gas regulator Z562483 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

control valve

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Press. Gas Compr. Gas - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service