Skip to Content
Merck
All Photos(1)

Key Documents

176710

Sigma-Aldrich

Deuterium bromide solution

47 wt. % in D2O, 99 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$403.00

$403.00


Estimated to ship onMay 20, 2025


Request more information

Select a Size

Change View
25 G
$403.00

About This Item

Linear Formula:
DBr/D2O
CAS Number:
Molecular Weight:
101.95
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.12

$403.00


Estimated to ship onMay 20, 2025


Request more information

isotopic purity

99 atom % D

form

liquid

concentration

47 wt. % in D2O

technique(s)

NMR: suitable

bp

126 °C (lit.)

density

1.537 g/mL at 25 °C

mass shift

M+1

SMILES string

Br[2H]

InChI

1S/BrH/h1H/i/hD

InChI key

CPELXLSAUQHCOX-DYCDLGHISA-N

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service