Skip to Content
Merck
All Photos(2)

Key Documents

484008

Sigma-Aldrich

Terbium(III) acetylacetonate hydrate

99.9% trace metals basis

Synonym(s):

2,4-Pentanedione terbium(III) derivative, Tb(acac)3, Terbium 2,4-pentanedionate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$84.30

$84.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$84.30

About This Item

Linear Formula:
Tb(C5H7O2)3 · xH2O
Molecular Weight:
456.25 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$84.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99.9% trace metals basis

form

powder or crystals

reaction suitability

core: terbium
reagent type: catalyst

mp

127 °C (dec.) (lit.)

SMILES string

O.CC(=O)\C=C(\C)O[Tb](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.H2O.Tb/c3*1-4(6)3-5(2)7;;/h3*3,6H,1-2H3;1H2;/q;;;;+3/p-3/b3*4-3-;;

InChI key

MRUSPMJZFLPYOU-KJVLTGTBSA-K

Application

Used in the preparation of a new family of monoporphyrinates interesting for their photochemical and magnetic properties and synthetic applications.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service