Skip to Content
Merck
All Photos(2)

Key Documents

325929

Sigma-Aldrich

Terbium(III) acetate hydrate

99.9% trace metals basis

Synonym(s):

Terbium triacetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$234.00
25 G
$820.00

$234.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$234.00
25 G
$820.00

About This Item

Linear Formula:
Tb(CH3CO2)3 · xH2O
CAS Number:
Molecular Weight:
336.06 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$234.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99.9% trace metals basis

form

crystals and lumps

reaction suitability

core: terbium
reagent type: catalyst

SMILES string

O.CC(=O)O[Tb](OC(C)=O)OC(C)=O

InChI

1S/3C2H4O2.H2O.Tb/c3*1-2(3)4;;/h3*1H3,(H,3,4);1H2;/q;;;;+3/p-3

InChI key

CUFVJHPGFMTTMB-UHFFFAOYSA-K

General description

Terbium(III) acetate hydrate is a crystalline compound with a high boiling and melting point. Due to its luminescent properties, it is mainly used as a precursor to produce phosphors, scintillators, catalysts, and magneto-optic materials.

Application

Terbium(III) acetate hydrate can be used:
  • As a precursor to prepare terbium oxide catalyst.[1]
  • As a surface passivator to fabricate photoluminescent carbon quantum dots.[2]
  • As a dopant to prepare Li co-doped ZnO nanoparticles which can be applied in electro-optic and magnetic devices.[3]
  • As a sol-gel precursor to prepare lead zirconate titanate thin films.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal genesis, characterization, and electrical conductivity measurements of terbium oxide catalyst obtained from terbium acetate
S.A. Soliman and B.M. Abu-Zied
Thermochimica Acta, 491, 84-91 (2009)
Ferroelectric properties of lanthanide-doped Pb (Zr 0.6, Ti 0.4) O3 thin films prepared by using a sol-gel method
Young-Hoon Son, et al.
Journal of Vacuum Science & Technology. A, Vacuum, Surfaces, And Films, 22, 1743-1745 (2004)
Lingling Xu et al.
Biomaterials, 230, 119670-119670 (2019-12-16)
Two-dimensional (2D) ultrathin nanomaterials have shown extensive attention and potential biomedical applications in cancer theranostics. Herein, for the first time, we report the synthesis of monodisperse ultrathin lanthanum oxyiodide (LaOI) nanosheets with a thickness of merely 3 nm based on a
Noah J J Johnson et al.
ACS nano, 10(9), 8299-8307 (2016-09-03)
Paramagnetic gadolinium (Gd(3+))-based nanocrystals (NCs) with a large number of confined gadolinium ions can be expected to heavily enhance the longitudinal (T1) relaxation of water protons compared to clinical gadolinium complexes with only a single paramagnetic center. However, paramagnetic Gd(3+)-NCs
Optical and magnetic properties of terbium doped zinc oxide nanoparticles with lithium as charge compensator
Pawan Kumar, et al.
Optik, 216, 164839-164839 (2020)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service