Skip to Content
Merck
All Photos(1)

Key Documents

483176

Sigma-Aldrich

Methylphenylsilane

95%

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$103.00

$103.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$103.00

About This Item

Linear Formula:
C6H5SiH2(CH3)
CAS Number:
Molecular Weight:
122.24
Beilstein:
2935287
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$103.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

refractive index

n20/D 1.506 (lit.)

bp

140-143 °C/751 mmHg (lit.)

density

0.89 g/mL at 25 °C (lit.)

SMILES string

[H][Si]([H])(C)c1ccccc1

InChI

1S/C7H10Si/c1-8-7-5-3-2-4-6-7/h2-6H,8H2,1H3

InChI key

QRLBICHXRCOJDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Slawomir Rubinsztajn et al.
Macromolecular rapid communications, 42(5), e2000601-e2000601 (2020-12-04)
The reaction of poly(hydromethylsiloxane-co-methylphenylsiloxane) with zirconium (IV) n-propoxide in dry toluene leads to extensive scission of the siloxane chain and conversion of Si-H groups. These processes produce oligomeric siloxanes and organosilanes containing moisture sensitive propoxy and siloxy-zirconate groups. The obtained
Hossein Mardani et al.
Molecules (Basel, Switzerland), 24(9) (2019-05-11)
Safranal, the main volatile chemical of Saffron (Crocus sativus) was studied to estimate its allelopathic effects on the photosynthetic pigment chlorophyll, leaf electrolyte leakage, fresh weight, catalase (CAT), and peroxidase (POX) activity of the test plant Lettuce (Lactuca sativa). In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service