Skip to Content
Merck
All Photos(1)

Key Documents

481645

Sigma-Aldrich

Poly[methyl(3,3,3-trifluoropropyl)siloxane]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H7F3OSi)n
CAS Number:
MDL number:
UNSPSC Code:
12352300

refractive index

n20/D 1.383 (lit.)

viscosity

10,000 cSt(25 °C)(lit.)

density

1.3 g/mL at 25 °C (lit.)

SMILES string

C[Si](=O)CCC(F)(F)F

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in glass bottles

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service