Skip to Content
Merck
All Photos(1)

Key Documents

476544

Sigma-Aldrich

PSS-Octakis(dimethylsilyloxy) substituted

Synonym(s):

1,3,5,7,9,11,13,15-Octakis(dimethylsilyloxy)pentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane, Octakis(dimethylsilyloxy)silsesquioxane

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$355.00

$355.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$355.00

About This Item

Empirical Formula (Hill Notation):
C16H56O20Si16
CAS Number:
Molecular Weight:
1017.97
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$355.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

mp

250 °C (dec.) (lit.)

SMILES string

[H][Si](C)(C)O[Si]12O[Si]3(O[Si]([H])(C)C)O[Si]4(O[Si]([H])(C)C)O[Si](O[Si]([H])(C)C)(O1)O[Si]5(O[Si]([H])(C)C)O[Si](O[Si]([H])(C)C)(O2)O[Si](O[Si]([H])(C)C)(O3)O[Si](O[Si]([H])(C)C)(O4)O5

InChI

1S/C16H56O20Si16/c1-37(2)17-45-25-46(18-38(3)4)28-49(21-41(9)10)30-47(26-45,19-39(5)6)32-51(23-43(13)14)33-48(27-45,20-40(7)8)31-50(29-46,22-42(11)12)35-52(34-49,36-51)24-44(15)16/h37-44H,1-16H3

InChI key

IZXLUEMTVRPVBL-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service