Skip to Content
Merck
All Photos(1)

Documents

468592

Sigma-Aldrich

PSS-Allyl-Heptacyclopentyl substituted

Synonym(s):

1-Allyl-3,5,7,9,11,13,15-heptacyclopentylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane, Allyl-POSS®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H68O12Si8
CAS Number:
Molecular Weight:
941.62
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

SMILES string

C=CC[Si]12O[Si]3(O[Si]4(O[Si](O1)(O[Si]5(O[Si](O2)(O[Si](O3)(O[Si](O4)(O5)C6CCCC6)C7CCCC7)C8CCCC8)C9CCCC9)C%10CCCC%10)C%11CCCC%11)C%12CCCC%12

InChI

1S/C38H68O12Si8/c1-2-31-51-39-52(32-17-3-4-18-32)42-55(35-23-9-10-24-35)44-53(40-51,33-19-5-6-20-33)46-57(37-27-13-14-28-37)47-54(41-51,34-21-7-8-22-34)45-56(43-52,36-25-11-12-26-36)49-58(48-55,50-57)38-29-15-16-30-38/h2,32-38H,1,3-31H2

InChI key

WPWQPIMCUOEXGC-UHFFFAOYSA-N

Application

Polyhedral oligomeric silsesquioxane (POSS) monomer, useful as model for silica surfaces, and for the formation of novel polymeric materials.

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Macromolecules, 28, 8435-8435 (1995)
Journal of the American Chemical Society, 112, 1931-1931 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service