Skip to Content
Merck
All Photos(3)

Key Documents

772046

Sigma-Aldrich

Fmoc-D-His(Trt)-OH

97% (HPLC)

Synonym(s):

Fmoc-N(im)-trityl-D-histidine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$133.00
5 G
$171.00
25 G
$360.00

$133.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$133.00
5 G
$171.00
25 G
$360.00

About This Item

Empirical Formula (Hill Notation):
C40H33N3O4
CAS Number:
Molecular Weight:
619.71
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

$133.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97% (HPLC)

form

powder or crystals

optical activity

[α]22/D -85.0°, c = 1% in chloroform

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

141-146 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H](Cc1cn(cn1)C(c2ccccc2)(c3ccccc3)c4ccccc4)NC(=O)OCC5c6ccccc6-c7ccccc57

InChI

1S/C40H33N3O4/c44-38(45)37(42-39(46)47-26-36-34-22-12-10-20-32(34)33-21-11-13-23-35(33)36)24-31-25-43(27-41-31)40(28-14-4-1-5-15-28,29-16-6-2-7-17-29)30-18-8-3-9-19-30/h1-23,25,27,36-37H,24,26H2,(H,42,46)(H,44,45)/t37-/m1/s1

InChI key

XXMYDXUIZKNHDT-DIPNUNPCSA-N

related product

Product No.
Description
Pricing

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seon-Yeong Kwak et al.
Amino acids, 46(12), 2777-2785 (2014-09-15)
Caffeic acid (CA) is one of the antioxidants found in plants, which protects vascular cells against vascular injuries from oxidative stress. In our previous study, caffeoyl-prolyl-histidine amide (CA-L-Pro-L-His-NH2; CA-PH; a CA derivative) was synthesized, which exhibited a strong antioxidant activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service