Select a Size
Select a Size
About This Item
Recommended Products
Assay
95%
mp
>350 °C (lit.)
functional group
bromo
SMILES string
Nc1nc(Br)c2nc[nH]c2n1
InChI
1S/C5H4BrN5/c6-3-2-4(9-1-8-2)11-5(7)10-3/h1H,(H3,7,8,9,10,11)
InChI key
HPGBGNVPUMCKPM-UHFFFAOYSA-N
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service