Skip to Content
Merck
All Photos(2)

Key Documents

427187

Sigma-Aldrich

Poly(vinylidene fluoride-co-hexafluoropropylene)

pellets

Synonym(s):

PVDF-HFP

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$94.60

$94.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$94.60

About This Item

Linear Formula:
(-CH2CF2-)x[-CF2CF(CF3)-]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$94.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

pellets

Quality Level

melt index

4-10 g/10 min (230°C/12.5kg)

dielectric constant

11.38, 100 Hz (ASTM D 750)

refractive index

n20/D 1.407

viscosity

20,000-25,000 poise(230 °C) (100 sec-1)(lit.)

transition temp

Tm 135-140 °C (ASTM D 3418)

density

1.78 g/mL at 25 °C

SMILES string

FC(F)=C.FC(F)=C(F)C(F)(F)F

InChI key

OQMIRQSWHKCKNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Wire and cable coatings, flexible and corrosion-resistant tubing, and liners for pipes and tanks.

Features and Benefits

Excellent thermal stability, chemical and abrasion resistance, impervious to UV degradation, self-extinguishing and retains properties on aging. Improved flexibility at subzero temperatures, stress crack resistance and elongation to break over poly(vinylidene fluoride).

Physical form

Crystalline, thermoplastic copolymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. I would like to purchase PVDF-HFP, may i know how should i know the content percentage of HFP?

    1 answer
    1. The HFP content is not tested on a lot-to lot-basis. Historical data suggest a range of 5-20%.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service