Skip to Content
Merck
All Photos(2)

Key Documents

463353

Sigma-Aldrich

tert-Butyl methacrylate

98%, contains 200 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

1,1-Dimethylethyl methacrylate, tert -Butyl α-methacrylate, tert -Butyl methacrylate, Acryester TB

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$108.00
250 ML
$217.00

$108.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$108.00
250 ML
$217.00

About This Item

Linear Formula:
H2C=C(CH3)CO2C(CH3)3
CAS Number:
Molecular Weight:
142.20
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$108.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Quality Level

Assay

98%

contains

200 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.415 (lit.)

bp

132 °C (lit.)

density

0.875 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OC(C)(C)C

InChI

1S/C8H14O2/c1-6(2)7(9)10-8(3,4)5/h1H2,2-5H3

InChI key

SJMYWORNLPSJQO-UHFFFAOYSA-N

Application

tert-Butyl methacrylate (tert-BMA) can be used in the formation of homo and block copolymers by atom transfer radical polymerization (ATRP) for potential usage in coatings, biomaterials and flocculants.[1][2][3]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Homo and block copolymers of tert-butyl methacrylate by atom transfer radical polymerization
Krishnan R and Srinivasan KSV
European Polymer Journal, 40(10), 2269-2276 (2004)
Termination Kinetics of tert-Butyl Methacrylate and of n-Butyl Methacrylate Free-Radical Bulk Homopolymerizations
Buback M and Junkers T
Macromolecular Chemistry and Physics, 207(18), 1640-1650 (2006)
Phase behavior of linear polystyrene-block-poly (2-vinylpyridine)-block-poly (tert-butyl methacrylate) triblock terpolymers
Ludwigs S, et al.
Polymer, 44(22), 6815-6823 (2003)
Hironori Kumagai et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 136, 29-37 (2019-01-15)
We are investigating an imaging agent for early detection of colorectal cancer. The agent, named the nanobeacon, is coumarin 6-encapsulated polystyrene nanospheres whose surfaces are covered with poly(N-vinylacetamide) and peanut agglutinin that reduces non-specific interactions with the normal mucosa and
Yue Liu et al.
Colloids and surfaces. B, Biointerfaces, 136, 721-728 (2015-10-30)
A novel nanoparticle with multilayer core-shell architecture for cell imaging is designed and synthesized by coating a fluorescent YVO4:Eu core with a diblock copolymer, MPEG-b-PMAA. The synthesis of YVO4:Eu core, which further makes MPEG-b-PMAA-YVO4:Eu NPs adapt for cell imaging, is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service