Skip to Content
Merck
All Photos(1)

Key Documents

434620

Sigma-Aldrich

Vinyl methacrylate

98%

Synonym(s):

Vinyl methacrylate

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$57.30

$57.30


Estimated to ship onMarch 27, 2025


Request a Bulk Order

Select a Size

Change View
25 ML
$57.30

About This Item

Linear Formula:
H2C=C(CH3)CO2CH=CH2
CAS Number:
Molecular Weight:
112.13
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$57.30


Estimated to ship onMarch 27, 2025


Request a Bulk Order

Quality Level

Assay

98%

form

liquid

contains

150-300 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.436 (lit.)

bp

111-112 °C (lit.)

density

0.933 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OC=C

InChI

1S/C6H8O2/c1-4-8-6(7)5(2)3/h4H,1-2H2,3H3

InChI key

FFYWKOUKJFCBAM-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengna Wang et al.
International journal of biological macromolecules, 166, 771-777 (2020-11-07)
A novel polymer poly (6-O-MMAGlc) has been synthesized via free radical polymerization of monomer methyl 6-O-methacryloyl-α-D-glucoside (6-O-MMAGlc) and characterized. The influence of poly(6-O-MMAGlc) on the formation of hen egg white lysozyme (HEWL) amyloid fibril was detailly investigated, indicating that the
Elizabeth M Boazak et al.
PloS one, 14(5), e0215895-e0215895 (2019-05-10)
The use of hydrogels in load bearing applications is often limited by insufficient toughness. 2-Hydroxyethyl methacrylate (HEMA) based hydrogels are appealing for translational work, as they are affordable and the use of HEMA is FDA approved. Furthermore, HEMA is photopolymerizable
Jérémie Brand et al.
Carbohydrate polymers, 169, 189-197 (2017-05-16)
Herein we propose a versatile method for the surface tailoring of cellulose nanocrystals (CNCs) based on the reactivity of vinyl ester molecules toward the accessible hydroxyl groups located at the surface of the nanoparticles. CNCs produced from wood pulp were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service