Skip to Content
Merck
All Photos(2)

Key Documents

461229

Sigma-Aldrich

2-Benzoylcyclohexanone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COC6H9(=O)
CAS Number:
Molecular Weight:
202.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

88-91 °C (lit.)

functional group

ketone
phenyl

SMILES string

O=C1CCCCC1C(=O)c2ccccc2

InChI

1S/C13H14O2/c14-12-9-5-4-8-11(12)13(15)10-6-2-1-3-7-10/h1-3,6-7,11H,4-5,8-9H2

InChI key

YTVQIZRDLKWECQ-UHFFFAOYSA-N

General description

2-Benzoylcyclohexanone can be prepared from the reaction between silyl enol ether of cyclohexanone and benzoyl fluoride.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The regioselective preparation of 1, 3-diketones within a micro reactor.
Wiles C, et al.
Chemical Communications (Cambridge, England), 10, 1034-1035 (2002)
Gilbert E Tumambac et al.
Chirality, 17(4), 171-176 (2005-04-13)
The kinetics of the racemization of 2-benzoylcyclohexanone 1 in hexanes, ethanol, and mixtures thereof have been investigated by time dependence of enantiomeric purity using enantioselective HPLC. In pure hexanes and ethanol, the racemization half-lives were determined as 552 and 23.8

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service