Skip to Content
Merck
All Photos(2)

Key Documents

806420

Sigma-Aldrich

3,5-Bis(trifluoromethyl)anilino(oxo)acetic acid

Synonym(s):

[(3,5-Bis(trifluoromethyl)phenyl)amino](oxo)acetic acid, [(3,5-Bis(trifluoromethyl)phenyl)carbamoyl]formic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$176.00

$176.00


Estimated to ship onApril 04, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$176.00

About This Item

Empirical Formula (Hill Notation):
C10H5F6NO3
Molecular Weight:
301.14
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

$176.00


Estimated to ship onApril 04, 2025


Request a Bulk Order

Assay

95%

Quality Level

form

powder

SMILES string

O=C(C(O)=O)NC1=CC(C(F)(F)F)=CC(C(F)(F)F)=C1

InChI

1S/C10H5F6NO3/c11-9(12,13)4-1-5(10(14,15)16)3-6(2-4)17-7(18)8(19)20/h1-3H,(H,17,18)(H,19,20)

InChI key

ZVYMTPNGDSQVIS-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

KitAlysis™ Cu C-N (Buchwald-Hartwig) cross-coupling high-throughput screening kit. Detailed Set-Up User Guide and a downloadable excel file for calculations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service