Skip to Content
Merck
All Photos(2)

Key Documents

461024

Sigma-Aldrich

Lanthanum(III) oxalate hydrate

99.99% trace metals basis

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$92.50

$92.50


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$92.50

About This Item

Linear Formula:
La2(C2O4)3 · xH2O
CAS Number:
Molecular Weight:
541.87 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

$92.50


Estimated to ship onMay 24, 2025


Request a Bulk Order

grade

for analytical purposes

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: lanthanum

SMILES string

O.O=C1O[La](OC1=O)OC(=O)C(=O)O[La]2OC(=O)C(=O)O2

InChI

1S/3C2H2O4.2La.H2O/c3*3-1(4)2(5)6;;;/h3*(H,3,4)(H,5,6);;;1H2/q;;;2*+3;/p-6

InChI key

KHZAMZAEBBFMFS-UHFFFAOYSA-H

Application

Reagent in a new preparation of single crystal niobium phosphide via a La-Nb-P-O complex intermediate.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Xu et al.
Inorganic chemistry, 35(4), 845-849 (1996-02-14)
Large single crystals of NbP have been prepared. A single-crystal X-ray diffraction study shows that it crystallizes in tetragonal symmetry with space group I4(1)md (No. 109) and lattice parameters a = 3.3324(2) Å, c = 11.3705(7) Å, and Z =

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service