Skip to Content
Merck
All Photos(5)

Key Documents

459925

Sigma-Aldrich

Platinum(IV) oxide

greener alternative

Synonym(s):

Adam’s catalyst, NSC 402624, Platinic oxide, Platinum dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$119.00
1 G
$330.00
5 G
$961.00

$119.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$119.00
1 G
$330.00
5 G
$961.00

About This Item

Linear Formula:
PtO2
CAS Number:
Molecular Weight:
227.08
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$119.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

composition

Pt, 79-83%

reaction suitability

core: platinum
reagent type: catalyst

greener alternative product characteristics

Designing Safer Chemicals
Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

≥75 m2/g

impurities

iron, low

mp

450 °C (lit.)

greener alternative category

SMILES string

O=[Pt]=O

InChI

1S/2O.Pt

InChI key

YKIOKAURTKXMSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Prepared by Adams′ nitrate fusion method.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Catalyst for hydrogenation of various functional groups; used as dehydrogenation catalyst; used for selective oxidation of primary alcohols.
Catalyst for:
  • Hydrohydrazination reactions
  • Oxidation of sugars
  • Low-temperature oxidation of carbon monoxide
  • Oxygen reduction reaction
  • Hydrogenation reactions
  • Hydrogenolysis of the cyclopropyl group into an isopropyl group
  • Hydroxycyclopropanation reactions

Pictograms

Flame over circle

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of GSKDevelopment of GSK's reagent guides ? embedding sustainability into reagent selections reagent guides ? embedding sustainability into reagent selection.
Adams JP, et al.
Green Chemistry, 15, 1542-1549 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service