Skip to Content
Merck
All Photos(2)

Key Documents

229040

Sigma-Aldrich

Platinum(IV) oxide monohydrate

99.9% trace metals basis

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$435.00

$435.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$435.00

About This Item

Linear Formula:
PtO2·H2O
CAS Number:
Molecular Weight:
245.09
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.23

$435.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99.9% trace metals basis

form

powder

composition

Pt, 78-80%

reaction suitability

reagent type: catalyst
core: platinum

SMILES string

O.O=[Pt]=O

InChI

1S/H2O.2O.Pt/h1H2;;;

InChI key

SPGAMGILENUIOF-UHFFFAOYSA-N

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service