Skip to Content
Merck
All Photos(1)

Documents

458589

Sigma-Aldrich

Isonicotinic anhydride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2O3
CAS Number:
Molecular Weight:
228.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

113-116 °C (lit.)

SMILES string

O=C(OC(=O)c1ccncc1)c2ccncc2

InChI

1S/C12H8N2O3/c15-11(9-1-5-13-6-2-9)17-12(16)10-3-7-14-8-4-10/h1-8H

InChI key

ORBIGZZHMAJXAZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of Nicotinic and Isonicotinic Anhydride.
Schrecker AW and Maury PB.
Journal of the American Chemical Society, 76(22), 5803-5803 (1954)
François Darrouzain et al.
Journal of pharmaceutical and biomedical analysis, 41(1), 228-232 (2006-01-13)
Two hydrophobic parameters (logkw-C18 and logkw-IAM, respectively) of a huperzine A series were extrapolated by high performance liquid chromatography (HPLC) using both C18 and immobilised artificial membrane (IAM) columns. A mathematical correlation between C18 and IAM hydrophobic parameters was completed
New synthesis of the antitumor 6H-pyrido [4, 3-b] carbazole alkaloid ellipticine.
Kano S, et al.
The Journal of Organic Chemistry, 46(14), 2979-2981 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service