Skip to Content
Merck
All Photos(1)

Key Documents

406465

Sigma-Aldrich

3-(2-Carboxyphenyl)propionic acid

99%

Synonym(s):

2-Carboxybenzenepropanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$180.00

$180.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$180.00

About This Item

Linear Formula:
HO2CC6H4CH2CH2CO2H
CAS Number:
Molecular Weight:
194.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$180.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

99%

form

solid

mp

166-168 °C (lit.)

SMILES string

OC(=O)CCc1ccccc1C(O)=O

InChI

1S/C10H10O4/c11-9(12)6-5-7-3-1-2-4-8(7)10(13)14/h1-4H,5-6H2,(H,11,12)(H,13,14)

InChI key

VEEXBQLWMFMATJ-UHFFFAOYSA-N

General description

3-(2-Carboxyphenyl)propionic acid (2-(2-carboxyethyl)benzoic acid) is an aromatic acid that undergoes cyclization to generate 1-indanone on heating. It is formed during the reaction of β-tetralone with KO2 in toluene.[1]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Superoxide oxidation: a novel route to aromatic 1, 2-dicarboxylic acids.
Sotiriou C, et al.
The Journal of Organic Chemistry, 55(7), 2159-2164 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service