Skip to Content
Merck
All Photos(3)

Key Documents

456802

Sigma-Aldrich

4-(Methylthio)phenylboronic acid

≥95%

Synonym(s):

4-(Methylsulfanyl)benzeneboronic acid, 4-(Methylsulfanyl)phenylboronic acid, 4-(Methylthio)benzeneboronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$89.90

$89.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$89.90

About This Item

Linear Formula:
CH3SC6H4B(OH)2
CAS Number:
Molecular Weight:
168.02
Beilstein:
3247219
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$89.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder

impurities

<15% anhydride

mp

210-214 °C (lit.)

functional group

thioether

SMILES string

CSc1ccc(cc1)B(O)O

InChI

1S/C7H9BO2S/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5,9-10H,1H3

InChI key

IVUHTLFKBDDICS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Suzuki-Miyaura cross coupling reactions with 2-aryl-1-bromo-1-nitroethenes or alkynyl bromides
  • Addition reactions with naphthyridine N-oxides
  • Oxyarylation of Heck reaction intermediates for synthesis of THF derivatives
  • Sulfoxidation reactions
  • Copper-catalyzed halogenation

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service