Skip to Content
Merck
All Photos(2)

Documents

447862

Sigma-Aldrich

Bis(butylcyclopentadienyl)zirconium(IV) dichloride

97%

Synonym(s):

Dibutylzirconocene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26Cl2Zr
CAS Number:
Molecular Weight:
404.53
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

core: zirconium
reagent type: catalyst
reaction type: Olefin Metathesis

mp

98.7-99.4 °C (lit.)

SMILES string

Cl[Zr]Cl.CCCC[C]1[CH][CH][CH][CH]1.CCCC[C]2[CH][CH][CH][CH]2

InChI

1S/2C9H13.2ClH.Zr/c2*1-2-3-6-9-7-4-5-8-9;;;/h2*4-5,7-8H,2-3,6H2,1H3;2*1H;/q;;;;+2/p-2

InChI key

KZUKCLOWAMFDDB-UHFFFAOYSA-L

Related Categories

Application

Catalyst for:
  • Copolymerization of ethylene-alpha-olefin
  • Ethylene polymerization

Metallocene supported catalyst

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service