Skip to Content
Merck
All Photos(1)

Key Documents

532126

Sigma-Aldrich

Dichloro[rac-ethylenebis(4,5,6,7-tetrahydro-1-indenyl)]zirconium(IV)

97%

Synonym(s):

Dichloro[rel-(7aR,7′aR)-1,2-ethanediylbis[(1,2,3,3a,7a-η)-4,5,6,7-tetrahydro-1H-inden-1-ylidene]]zirconium

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$530.00

$530.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$530.00

About This Item

Empirical Formula (Hill Notation):
C20H24Cl2Zr
CAS Number:
Molecular Weight:
426.53
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$530.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

reaction suitability

core: zirconium
reagent type: catalyst

mp

267-271 °C (lit.)

SMILES string

Cl[Zr]Cl.[CH]1[CH][C](CC[C]2[CH][CH][C]3CCCC[C]23)[C]4CCCC[C]14

InChI

1S/C20H24.2ClH.Zr/c1-3-7-19-15(5-1)9-11-17(19)13-14-18-12-10-16-6-2-4-8-20(16)18;;;/h9-12H,1-8,13-14H2;2*1H;/q;;;+2/p-2

InChI key

AKXWJOYQORUUHS-UHFFFAOYSA-L

Application

Catalyst for:
  • Regiochemistry in Negishi carboaluminations
  • Polymerization of olefins
  • Diastereoselective intramolecular olefin alkylations
  • Asymmetric carbomagnesiation
  • Reduction of esters to alcohols
  • Enantioselective synthesis of allylic amines

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service