Skip to Content
Merck
All Photos(1)

Key Documents

446777

Sigma-Aldrich

Perfluoroundecanoic acid

95%

Synonym(s):

Heneicosafluoroundecanoic acid, PFUnDA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$128.00

$128.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$128.00

About This Item

Linear Formula:
CF3(CF2)9CO2H
CAS Number:
Molecular Weight:
564.09
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

$128.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

solid

bp

160 °C/60 mmHg (lit.)

mp

96-101 °C (lit.)

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C11HF21O2/c12-2(13,1(33)34)3(14,15)4(16,17)5(18,19)6(20,21)7(22,23)8(24,25)9(26,27)10(28,29)11(30,31)32/h(H,33,34)

InChI key

SIDINRCMMRKXGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Clara et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 58(1), 59-66 (2008-07-26)
Effluents of wastewater treatment plants are relevant point sources for the emission of hazardous xenobiotic substances to the aquatic environment. One group of substances, which recently entered scientific and political discussions, is the group of the perfluorinated alkylated substances (PFAS).
Shuhong Fang et al.
Environmental toxicology and chemistry, 35(12), 3005-3013 (2016-05-11)
Carp (Cyprinus carpio) were exposed to perfluoroalkyl acids (PFAAs) including perfluorooctane sulfonate (PFOS) isomers in an artificially contaminated sediment/water microcosm. The uptake constant of PFAAs increased with increasing carbon chain length, whereas the elimination coefficient displayed the opposite trend, suggesting
Po Wang et al.
Chemosphere, 263, 128325-128325 (2020-12-11)
The application of the organic-diffusive gradients in thin films (o-DGT) passive sampling technique for the monitoring of per- and polyfluoroalkyl substances (PFAS) in the environment is still limited. Six common PFAS with different chain lengths were evaluated in water by
Eugenia Pasanisi et al.
The Science of the total environment, 572, 1059-1065 (2016-08-16)
Lepidochelys olivacea is the most abundant and globally distributed sea turtle species in the world and thus, monitoring this species for persistent organic pollutants, such as perfluorinated chemicals, is fundamental for their protection. This study was the first to evaluate
Jonathan E Naile et al.
Chemosphere, 144, 1722-1728 (2015-11-01)
Perfluoroalkyl substances are globally distributed in both urban and remote settings, and routinely are detected in wildlife, humans, and the environment. One of the most prominent and routinely detected perfluoroalkyl substances is perfluorooctanoic acid (PFOA), which has been shown to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service