Skip to Content
Merck
All Photos(1)

Key Documents

443980

Sigma-Aldrich

2,4,6-Tris(trifluoromethyl)-1,3,5-triazine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$288.00

$288.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$288.00

About This Item

Empirical Formula (Hill Notation):
C6F9N3
CAS Number:
Molecular Weight:
285.07
Beilstein:
302339
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$288.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.313 (lit.)

bp

98.3-98.5 °C/748 mmHg (lit.)

density

1.596 g/mL at 25 °C

functional group

fluoro

SMILES string

FC(F)(F)c1nc(nc(n1)C(F)(F)F)C(F)(F)F

InChI

1S/C6F9N3/c7-4(8,9)1-16-2(5(10,11)12)18-3(17-1)6(13,14)15

InChI key

LSGBKABSSSIRJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Reaction of 1,3-bis(trimethylsilyloxy)-1,3-butadienes with 2,4,6-tris(trifluoromethyl)-1,3,5-triazine: planning and serendipity.
Viktor O Iaroshenko et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(26), 7188-7192 (2011-05-14)
Analytical Chemistry, 47, 2289-2289 (1975)
Karin Kipper et al.
Journal of chromatography. A, 1218(45), 8175-8180 (2011-10-05)
Two fluoroalcohols--1,1,1,3,3,3-hexafluoro-2-propanol (HFIP) and 1,1,1,3,3,3-hexafluoro-2-methyl-2-propanol (HFTB)--were evaluated as volatile buffer acids in basic mobile phases for LC-ESI-MS determination of acidic and basic compounds. HFIP and HFTB as acidic buffer components offer interesting possibilities to adjust retention behavior of different analytes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service