Skip to Content
Merck
All Photos(1)

Key Documents

442895

Sigma-Aldrich

Mercury(I) sulfate

96%

Synonym(s):

Mercurous sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hg2SO4
CAS Number:
Molecular Weight:
497.24
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
96%
form:
powder

Assay

96%

form

powder

density

7.56 g/mL at 25 °C (lit.)

SMILES string

[Hg+].[Hg+].[O-]S([O-])(=O)=O

InChI

1S/2Hg.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

MINVSWONZWKMDC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Somaye Rasouli et al.
Nano letters, 19(1), 46-53 (2018-12-07)
The evolution of Pt-Ni nanoparticles supported on amorphous carbon is investigated before and after electrochemical potential cycling (0.6-1.1 V), using aberration-corrected scanning transmission electron microscopy (STEM) and energy dispersive X-ray spectroscopy (EDS). During voltage cycling and due to the dissolution
Ellen E Benn et al.
Journal of the American Chemical Society, 139(10), 3663-3668 (2017-02-18)
Electroreduction of small molecules in aqueous solution often competes with the hydrogen evolution reaction (HER), especially if the reaction is driven even moderately hard using a large overpotential. Here, the oxygen reduction reaction (ORR) was studied under proton diffusion-limited conditions
Celestino Gomes et al.
Materials (Basel, Switzerland), 13(7) (2020-04-15)
This work investigated the use of ZnAl-layered double hydroxide (LDH) intercalated with nitrate or nitrite ions for controlling the corrosion of steel in reinforced concrete. The work started by analyzing the stability of the powder in the 1-14 pH range
Zhirong Sun et al.
Environmental science and pollution research international, 24(16), 14355-14364 (2017-04-22)
Palladium/carbon nanotubes-nafion film-modified titanium mesh electrode (Pd/CNTs-nafion film/Ti electrode) was prepared and used for catalytic dechlorination of 2,3,5-trichlorophenol (2,3,5-TCP). The influences of factors, such as Pd
Paul I Dargan et al.
Critical care (London, England), 7(3), R1-R6 (2003-06-10)
Inorganic mercury poisoning is uncommon, but when it occurs it can result in severe, life-threatening features and acute renal failure. Previous reports on the use of extracorporeal procedures such as haemodialysis and haemoperfusion have shown no significant removal of mercury.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service