Skip to Content
Merck
All Photos(1)

Key Documents

442666

Sigma-Aldrich

(+)-(18-Crown-6)-2,3,11,12-tetracarboxylic acid

97%

Synonym(s):

(2R,3R,11R,12R)-1,4,7,10,13,16-Hexaoxacyclooctadecane-2,3,11,12-tetracarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24O14
CAS Number:
Molecular Weight:
440.35
Beilstein:
4240375
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D +62°, c = 1 in methanol

mp

210-212 °C (lit.)

functional group

carboxylic acid
ether

SMILES string

OC(=O)[C@@H]1OCCOCCO[C@H]([C@@H](OCCOCCO[C@H]1C(O)=O)C(O)=O)C(O)=O

InChI

1S/C16H24O14/c17-13(18)9-10(14(19)20)29-7-3-26-4-8-30-12(16(23)24)11(15(21)22)28-6-2-25-1-5-27-9/h9-12H,1-8H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24)/t9-,10-,11-,12-/m1/s1

InChI key

FZERLKNAJSFDSQ-DDHJBXDOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Derived tetracarboxylic acid chloride of this crown ether reacts with meta- and para-xylylene diamines to form chiral captands in high yields. Used as a chiral selector in capillary zone electrophoresis for the separation of a variety of optically active amines. Synthesis of monoamides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzanne Anouti et al.
Electrophoresis, 31(6), 1029-1035 (2010-03-24)
The use of transient moving chemical reaction boundary (tMCRB) was investigated for the on-line preconcentration of native amino acids in heart-cutting 2D-CE with multiple detection points using contactless conductivity detection. The tMCRB focusing was obtained by using ammonium formate (pH
Su Jin Lee et al.
Journal of chromatography. A, 1188(2), 318-321 (2008-03-19)
Racemic vigabatrin, an anticonvulsant drug used for the treatment of epilepsy, and its analogue gamma-amino acids were resolved without derivatization on liquid chromatographic chiral stationary phases (CSPs) based on (+)-(18-crown-6)-2,3,11,12-tetracarboxylic acid. Between the two CSPs which contain 3 methylene-unit or
Taeho Lee et al.
Journal of chromatography. A, 1217(8), 1425-1428 (2010-01-12)
(-)-(18-Crown-6)-2,3,11,12-tetracarboxylic acid-bonded silica was used as the chiral stationary phase in capillary electrochromatography (CEC) for enantioseparation of some alpha-amino acids. Separation data in CEC were measured in mobile phases of varying pH, and composition of methanol and buffer, and compared
Je Young Park et al.
Chirality, 24(5), 427-431 (2012-04-18)
3-Amino-5-phenyl (or 5-methyl)-1,4-benzodiazepin-2-ones, which are chiral precursors of anti-respiratory syncytial virus active agents, were resolved on three different chiral stationary phases (CSPs) based on (+)-(18-crown-6)-2,3,11,12-tetracarboxylic acid or (3,3'-diphenyl-1,1'-binaphthyl)-20-crown-6. Among the three CSPs, the CSP that is based on (3,3'-diphenyl-1,1'-binaphthyl)-20-crown-6 and
Suzanne Anouti et al.
Electrophoresis, 30(1), 2-10 (2008-12-25)
A new methodology based on heart-cutting 2-D CE in a single capillary was developed for the chiral separation of a mixture of 22 underivatized amino acids. The first dimension is performed in an achiral BGE (2.3 M acetic acid, pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service