Skip to Content
Merck
All Photos(1)

Key Documents

388424

Sigma-Aldrich

2-Hydroxymethyl-15-crown-5

95%

Synonym(s):

1,4,7,10,13-Pentaoxacyclopentadecane-2-methanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$126.00

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$126.00

About This Item

Empirical Formula (Hill Notation):
C11H22O6
CAS Number:
Molecular Weight:
250.29
Beilstein:
3589706
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

viscous liquid

refractive index

n20/D 1.479 (lit.)

bp

135 °C/1.5 mmHg (lit.)

density

1.175 g/mL at 25 °C (lit.)

functional group

ether
hydroxyl

SMILES string

OCC1COCCOCCOCCOCCO1

InChI

1S/C11H22O6/c12-9-11-10-16-6-5-14-2-1-13-3-4-15-7-8-17-11/h11-12H,1-10H2

InChI key

YHIQMMGCRYKJLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service