Skip to Content
Merck
All Photos(2)

Key Documents

441775

Sigma-Aldrich

3,9-Bis(octadecyloxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane

Synonym(s):

Cyclic neopentanetetraylbis(octadecyl phosphite), Dioctadecyl pentaerythritol diphosphite, Distearyl pentaerythritol diphosphite, Phosphorous acid, dicyclic ester with pentaerythritol, dioctadecyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$60.60

$60.60


Estimated to ship onMarch 29, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$60.60

About This Item

Empirical Formula (Hill Notation):
C41H82O6P2
CAS Number:
Molecular Weight:
733.03
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$60.60


Estimated to ship onMarch 29, 2025


Request a Bulk Order

form

solid

Quality Level

mp

44-47 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCOP1OCC2(CO1)COP(OCCCCCCCCCCCCCCCCCC)OC2

InChI

1S/C41H82O6P2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-42-48-44-37-41(38-45-48)39-46-49(47-40-41)43-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-40H2,1-2H3

InChI key

PZRWFKGUFWPFID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,9-Bis(octadecyloxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane is a diphosphaspiro compound which contains two phosphorus atoms and can be used as an antioxidant and as a reducing agent.[1][2][3][4]

Application

Color stabilizer for polymers

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microplastics as a vector of hydrophobic contaminants: Importance of hydrophobic additives
Kwon J, et al.
Integrated Environmental Assessment and Management, 13(3), 494-499 (2017)
Estimation of the biobased carbon content of polypropylene resin in composites on the basis of the carbon 14 concentration
Taguchi K, et al.
Journal of Applied Polymer Science, 131(6), 40-48 (2014)
The formulator's guide to anti-oxygen inhibition additives
Husar B, et al.
Progress in Organic Coatings, 77(11), 1789-1798 (2014)
From Laboratory to Industrial Continuous Production of Polylactic Acid with Low Residual Monomer
Macromolecular Symposia, 360(1), 40-48 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service