Skip to Content
Merck
All Photos(1)

Key Documents

440094

Sigma-Aldrich

5-Chloro-2-fluorotoluene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(Cl)F
CAS Number:
Molecular Weight:
144.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.4995 (lit.)

bp

157-159 °C/752 mmHg (lit.)

density

1.188 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(Cl)ccc1F

InChI

1S/C7H6ClF/c1-5-4-6(8)2-3-7(5)9/h2-4H,1H3

InChI key

JOXXHDGUTVUBDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dorus Heijnen et al.
Organic letters, 17(9), 2262-2265 (2015-04-15)
The palladium-catalyzed direct cross-coupling of a range of organic chlorides and bromides with the bifunctional C(sp(3))-(trimethylsilyl)methyllithium reagent is reported. The use of Pd-PEPPSI-IPent as the catalyst allows for the preparation of structurally diverse and synthetically versatile benzyl- and allylsilanes in
K L Shepard et al.
Journal of medicinal chemistry, 34(10), 3098-3105 (1991-10-01)
For several decades a tantalizing goal for the treatment of primary open-angle glaucoma has been the development of a topically active carbonic anhydrase inhibitor. Recent results from several research groups indicate that considerable progress has been made toward this objective.
H Kanno et al.
Chemical & pharmaceutical bulletin, 40(8), 2049-2054 (1992-08-01)
New 1,4-dihydropyridine derivatives bearing a 4-(disubstituted phenyl) ring and an aminoethyl ester or an amino-2,2-dimethyl-propyl ester were synthesized and their antihypertensive activities were examined in normotensive rats and spontaneously hypertensive rats. The effects of phenyl substituents and ester groups on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service