Skip to Content
Merck
All Photos(1)

Key Documents

779946

Sigma-Aldrich

1-Bromo-4-phenylbutane

≥95.0% (GC)

Synonym(s):

(4-Bromobutyl)benzene, 4-Phenylbutyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$43.60
10 G
$367.00

$43.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$43.60
10 G
$367.00

About This Item

Empirical Formula (Hill Notation):
C10H13Br
CAS Number:
Molecular Weight:
213.11
Beilstein:
2042124
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$43.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95.0% (GC)
95.0-105.0% (T)

form

liquid

functional group

bromo
phenyl

SMILES string

BrCCCCc1ccccc1

InChI

1S/C10H13Br/c11-9-5-4-8-10-6-2-1-3-7-10/h1-3,6-7H,4-5,8-9H2

InChI key

XPBQQAHIVODAIC-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service