Skip to Content
Merck
All Photos(1)

Key Documents

437018

Sigma-Aldrich

N-Boc-1,6-hexanediamine hydrochloride

97%

Synonym(s):

N-Boc-1,6-diamino-hexane hydrochloride, tert-Butyl N-(6-aminohexyl)carbamate hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$104.00
5 G
$276.00

$104.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$104.00
5 G
$276.00

About This Item

Linear Formula:
(CH3)3COCONH(CH2)6NH2 · HCl
CAS Number:
Molecular Weight:
252.78
Beilstein:
3631740
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$104.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

reaction suitability

reagent type: cross-linking reagent

mp

162-164 °C (lit.)

functional group

Boc
amine

SMILES string

NCCCCCCNC(OC(C)(C)C)=O.[H]Cl

InChI

1S/C11H24N2O2.ClH/c1-11(2,3)15-10(14)13-9-7-5-4-6-8-12;/h4-9,12H2,1-3H3,(H,13,14);1H

InChI key

JSBWQIZQJOQPFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Boc-1,6-hexanediamine hydrochloride is a general reagent to introduce a C6-spacer. It can be used in:
  • Synthesis of poly(L-glutamic acid) gadolinium chelates as biodegradable MRI contrast agents.[1]
  • Synthesis of cholesteryl-functionalized hyaluronic acid-based anionic nanogel for protein delivery.[2]
  • Preparation of amphiphilic prodrug, containing anticancer drug chlorambucil, which would self-assemble to form micelles in aqueous solution.[3]

Reagent for the introduction of a C6-spacer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rational Design of an Amphiphilic Chlorambucil Prodrug Realizing Self-Assembled Micelles for Efficient Anticancer Therapy.
Hu X, et al.
ACS biomaterials science & engineering, 4(3), 973-980 (2018)
Injectable hydrogel for sustained protein release by salt?induced association of hyaluronic acid nanogel.
Nakai T, et al.
Macromolecular Bioscience, 12(4), 475-483 (2012)
Synthesis and characterization of poly (L-glutamic acid) gadolinium chelate: a new biodegradable MRI contrast agent.
Wen X, et al.
Bioconjugate Chemistry, 15(6), 1408-1415 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service