Skip to Content
Merck
All Photos(2)

Key Documents

435848

Sigma-Aldrich

Indium(III) fluoride

≥99.9% trace metals basis

Synonym(s):

Trifluoroindigane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$68.40

$68.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$68.40

About This Item

Linear Formula:
InF3
CAS Number:
Molecular Weight:
171.81
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

$68.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: indium

impurities

≤1000.0 ppm Trace Metal Analysis

SMILES string

F[In](F)F

InChI

1S/3FH.In/h3*1H;/q;;;+3/p-3

InChI key

JNLSTWIBJFIVHZ-UHFFFAOYSA-K

General description

Indium(III) fluoride is a white crystalline solid with a relatively high melting point. It is a wide band gap semiconductor suitable for optoelectronic applications. Non-oxide glasses are made using indium(III) fluoride.

Application

Indium(III) fluoride can be used:
  • As a precursor to prepare indium oxide thin films for various applications.[1]
  • To fabricate Pr3+/Er3+ co-doped glasses for white light emission.[2]
  • For surface fluorination treatment of Indium-based quantum dots to enhance their photoluminescence quantum yield.[3]
  • As an electrolyte additive for Li-metal batteries to enhance theircycling performance.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

White light emission and energy transfer in Pr3+/Er3+ co-doped InF3-based glass
Joanna Pisarska, et al.
Materials Research Bulletin, 150, 111791-111791 (2022)
Eur. J. Solid State Inorg. Chem., 33, 879-879 (1996)
Trap Passivation in Indium-Based Quantum Dots through Surface Fluorination: Mechanism and Applications
Tae-Gon Kim, et al.
ACS Nano, 12, 11529-11540 (2018)
Nitrile Electrolyte Strategy for 4.9 V-Class Lithium-Metal Batteries Operating in Flame
Hyunseok Moon, et al.
Energy & Environmental Science, 6, e12383-e12383 (2023)
Loh T-P. et al.
Synlett, 369-369 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service