Skip to Content
Merck
All Photos(1)

Key Documents

433926

Sigma-Aldrich

1′,3′-Dihydro-8-methoxy-1′,3′,3′-trimethyl-6-nitrospiro[2H-1-benzopyran-2,2′-(2H)-indole]

97%

Synonym(s):

1′3′,3′-Trimethyl-6-nitro-8-methoxyspiro[2-1-benzopyran-2,2′-indoline], 1,3,3-Trimethyl-6′-nitro-8′-methoxyspiro(indoline-2,2′-benzopyran), 1,3,3-Trimethylindolino-6′-nitro-8′-methoxybenzopyrylspiran, 1,3,3-Trimethylindolino-8′-methoxy-6′-nitrobenzospiropyran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20N2O4
CAS Number:
Molecular Weight:
352.38
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

mp

159-162 °C (lit.)

SMILES string

COc1cc(cc2C=CC3(Oc12)N(C)c4ccccc4C3(C)C)[N+]([O-])=O

InChI

1S/C20H20N2O4/c1-19(2)15-7-5-6-8-16(15)21(3)20(19)10-9-13-11-14(22(23)24)12-17(25-4)18(13)26-20/h5-12H,1-4H3

InChI key

IZDVWQPIYXTGIF-UHFFFAOYSA-N

General description

1′,3′-Dihydro-8-methoxy-1′,3′,3′-trimethyl-6-nitrospiro[2H-1-benzopyran-2,2′-(2H)-indole] (6-NO2-8-OMe-BIPS) is a spiropyran based photochromic dye. It has a nitro group that enhances the overall quantum yield of the photochemical system.[1][2]

Application

6-NO2-8-OMe-BIPS can be potentially used in a variety of applications such as switching devices, signal transducer receptors, and optical imaging processes.[2][3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

UV-responsive polyvinyl alcohol nanofibers prepared by electrospinning.
Khatri Z, et al.
Applied Surface Science, 342, 64-68 (2015)
Spiropyran, chromene and spirooxazine, melange trois: Molecular logic systems through selective and reversible deactivation of photochromism mediated by CO2 gas.
Darwish TA, et al.
Dyes and Pigments, 92(2), 817-824 (2012)
Complexes of spiropyran-derived merocyanines with metal ions: relaxation kinetics, photochemistry and solvent effects.
Chibisov AK and Gorner H
Chemical Physics, 237(3), 425-442 (1998)

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service