Skip to Content
Merck
All Photos(1)

Key Documents

434086

Sigma-Aldrich

3-Methoxy-5-nitrosalicylaldehyde

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$242.00

$242.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$242.00

About This Item

Linear Formula:
CH3OC6H2(NO2)-2-(OH)CHO
CAS Number:
Molecular Weight:
197.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$242.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

141-143 °C (lit.)

functional group

aldehyde
nitro

SMILES string

COc1cc(cc(C=O)c1O)[N+]([O-])=O

InChI

1S/C8H7NO5/c1-14-7-3-6(9(12)13)2-5(4-10)8(7)11/h2-4,11H,1H3

InChI key

HGKHVFKBOHFYSS-UHFFFAOYSA-N

Application

3-Methoxy-5-nitrosalicylaldehyde may be used for the following syntheses:
  • zinc-selective spiropyran-based fluorescent and photoregenerable receptor[1]
  • (E)-2,4-dihydroxy-N′-(2-hydroxy-3-methoxy-5-nitrobenzylidene)benzohydrazide dihydrate[2]
  • ethyl 4-(8-methoxy-3′,3′-dimethyl-6-nitrospiro[chromene-2,2′-indoline]-1′-yl)butanoate[3]
  • 1-(3-carbomethoxypropyl)-3-3-dimethyl-8-methoxy-6-nitrospiro[2H-1]-benzopyran-2,2-indoline[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A photoswitchable Zn (II) selective spiropyran-based sensor.
Natali M, et al.
Tetrahedron, 66(38), 7612-7617 (2010)
The balance between closed and open forms of spiropyrans in the solid state.
Aakeroy CB, et al.
CrystEngComm, 12(4), 1027-1033 (2010)
You-Yue Han et al.
Acta crystallographica. Section E, Structure reports online, 66(Pt 5), o1085-o1086 (2010-01-01)
The asymmetric unit of the title compound, C(15)H(13)N(3)O(7)·2H(2)O, consists of a hydrazone mol-ecule and two solvent water mol-ecules. The mol-ecule adopts an E configuration with respect to the C=N bond. It is relatively planar, with a dihedral angle between the
Spiropyran-based reversible, light-modulated sensing with reduced photofatigue.
Radu A, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 206(2), 109-115 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service