Skip to Content
Merck
All Photos(1)

Documents

433799

Sigma-Aldrich

1-Decyl-2-methylimidazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H26N2
CAS Number:
Molecular Weight:
222.37
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.475 (lit.)

bp

157-158 °C/3 mmHg (lit.)

density

0.9 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCn1ccnc1C

InChI

1S/C14H26N2/c1-3-4-5-6-7-8-9-10-12-16-13-11-15-14(16)2/h11,13H,3-10,12H2,1-2H3

InChI key

BKFRZOZNMWIFLH-UHFFFAOYSA-N

General description

1-Decyl-2-methylimidazole is an 1-alkyl-2-methylimidazole. It participates in the extraction process of Cu(II) complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of the Steric Hindrance, Ligand Hydrophobicity, and DN of solvents on Structure and Extraction of Cu (II) Complexes of 1-Alkyl-2-Ethylimidazoles.
Radzyminska-Lenarcik E.
Separation Science and Technology, 43(4), 794-814 (2008)
The influence of alkyl chain length and steric effect on extraction of zinc (II) complexes with 1-alkyl-2-methylimidazoles.
Lenarcik B and Kierzkowska A.
Solvent Extr. Ion Exch., 24(3), 433-445 (2006)
CATALYSIS-APPLIED AND PHYSICAL ASPECTS.
Platinum Metals Rev., 56(4), 281-283 (2012)
David Valverde et al.
ChemSusChem, 12(17), 3996-4004 (2019-07-10)
The catalytic activity of Rose Bengal (RB) immobilized on supported ionic liquid (IL)-like phases was evaluated as a polymer-supported photocatalyst. In these systems, the polymer was designed to play a pivotal role. The polymeric backbone adequately modified with IL-like moieties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service