Skip to Content
Merck
All Photos(1)

Documents

424951

Sigma-Aldrich

Ethyl (S)-(−)-2-(tert-butyldimethylsilyloxy)propionate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH[OSi(CH3)2C(CH3)3]CO2C2H5
CAS Number:
Molecular Weight:
232.39
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

optical activity

[α]20/D −30°, c = 1.26 in chloroform

refractive index

n20/D 1.425 (lit.)

bp

187 °C (lit.)

density

0.875 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)[C@H](C)O[Si](C)(C)C(C)(C)C

InChI

1S/C11H24O3Si/c1-8-13-10(12)9(2)14-15(6,7)11(3,4)5/h9H,8H2,1-7H3/t9-/m0/s1

InChI key

KOBAXFIJEBLKRZ-VIFPVBQESA-N

Application

Precursor to the lactyl-3,4-didehydroproline didemnin B side chain. Starting material for the asymmetric synthesis of syn- and anti- functionalized1,2-diols.

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mayer, S.C. et al.
The Journal of Organic Chemistry, 59, 5192-5192 (1994)
Solladie, G. Almario, A.
Tetrahedron Letters, 35, 1937-1937 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service