Skip to Content
Merck
All Photos(1)

Key Documents

423823

Sigma-Aldrich

Tetraamminepalladium(II) nitrate solution

10 wt. % in H2O, 99.99%

Synonym(s):

Tetraamminedinitratopalladium, Tetraamminepalladium dinitrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$268.00

$268.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$268.00

About This Item

Linear Formula:
Pd(NH3)4(NO3)2
CAS Number:
Molecular Weight:
298.55
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$268.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99.99%

form

liquid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

concentration

10 wt. % in H2O

density

1.03 g/mL at 25 °C

SMILES string

N.N.N.N.[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.4H3N.Pd/c2*2-1(3)4;;;;;/h;;4*1H3;/q2*-1;;;;;+2

InChI key

WIDMMNCAAAYGKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetraamminepalladium(II)nitrate solution can be used as a palladium precursor to prepare:
  • Pd/C catalyst for the decarbonylation of heptanoic acid to 1-hexene.
  • Pd/Al2O3 catalyst for the decomposition of formic acid.
  • Pd/CNT catalyst for the dehalogenation of triclosan in water.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Berry
Journal of submicroscopic cytology, 19(1), 53-56 (1987-01-01)
Palladium salt has been used for some time in experimental therapy protocols; with this in mind, we carried out a study of the effect of tetramine dichloro-palladium (soluble salt) upon kidney cells. Using an electron microprobe, we were able to
J Ma
The Journal of general physiology, 102(6), 1031-1056 (1993-12-01)
The effects of ruthenium red and the related compounds tetraamine palladium (4APd) and tetraamine platinum (4APt) were studied on the ryanodine activated Ca2+ release channel reconstituted in planar bilayers with the immunoaffinity purified ryanodine receptor. Ruthenium red, applied at submicromolar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service