Skip to Content
Merck
Tetraamminepalladium(II) nitrate solution 10 wt. % in H2O, 99.99%

Tetraamminepalladium(II) nitrate solution

Tetraamminepalladium(II) nitrate solution 10 wt. % in H2O, 99.99%
Synonym(s):
Tetraamminedinitratopalladium, Tetraamminepalladium dinitrate
Linear Formula:
Pd(NH3)4(NO3)2
CAS Number:
Molecular Weight:
298.55

Products

Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Core