Skip to Content
Merck
All Photos(1)

Documents

42254

Sigma-Aldrich

1-Butyl-3-methylimidazolium thiocyanate

≥95%

Synonym(s):

1-Butyl-3-methyl-1H-imidazolium thiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N3S
CAS Number:
Molecular Weight:
197.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

liquid

impurities

≤1% water

SMILES string

[S-]C#N.CCCCn1cc[n+](C)c1

InChI

1S/C8H15N2.CHNS/c1-3-4-5-10-7-6-9(2)8-10;2-1-3/h6-8H,3-5H2,1-2H3;3H/q+1;/p-1

InChI key

SIXHYMZEOJSYQH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

1-Butyl-3-methylimidazolium thiocyanate ([BMIM][SCN]) ionic liquid can be used:
  • As an alternative to conventional entrainers in the extractive distillation of cyclohexane and benzene.
  • In polyethylene oxide-sodium perchlorate electrolyte to increase its ionic conductivity at room temperature.

[BMIM][SCN] show higher degree of solubility of lithium salts such as lithium perchlorate (LiClO4) and N-lithiotrifluoromethanesulfonimide(LiTFSI) when compared to other room temperature ionic liquids due to the presence of polarizable anion (-SCN).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

403.7 °F - closed cup

Flash Point(C)

206.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzene and cyclohexane separation using 1-butyl-3-methylimidazolium thiocyanate
Gonfa G, et al.
AIP Conference Proceedings, 1877(1), 020005-020005 (2017)
Solubility, ionic conductivity and viscosity of lithium salts in room temperature ionic liquids
Rosol ZP, et al.
Green Chemistry, 11(9), 1453-1457 (2009)
Effect of addition of 1-butyl-3-methylimidazolium thiocyanate on conductivity of Na-containing polymer electrolyte
Lee S, et al.
Research on Chemical Intermediates, 43(10), 5403-5411 (2017)
Daisuke Sugioka et al.
Physical chemistry chemical physics : PCCP, 17(19), 13150-13159 (2015-04-29)
Sputtering of noble metals, such as Au, Ag, Pd, and Pt, onto room-temperature ionic liquids (RTILs) enabled the formation of monoparticle films composed of spherical noble metal nanoparticles on the liquid surface only when the RTILs used contained hydroxyl-functionalized cations

Articles

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service