Skip to Content
Merck
All Photos(1)

Key Documents

420204

Sigma-Aldrich

2-Methyl-2-phenylpropylmagnesium chloride solution

0.5 M in diethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$162.00

$162.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$162.00

About This Item

Linear Formula:
C6H5C(CH3)2CH2MgCl
CAS Number:
Molecular Weight:
192.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$162.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in diethyl ether

bp

34.6 °C

density

0.766 g/mL at 25 °C

functional group

phenyl

SMILES string

CC(C)(C[Mg]Cl)c1ccccc1

InChI

1S/C10H13.ClH.Mg/c1-10(2,3)9-7-5-4-6-8-9;;/h4-8H,1H2,2-3H3;1H;/q;;+1/p-1

InChI key

BXKYFONHSSFLKR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.0 °F - closed cup

Flash Point(C)

-27.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service