419370
3,4-Dihydroxybenzaldehyde
synthesis grade
Synonym(s):
Protocatechualdehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
synthesis grade
mp
150-157 °C (lit.)
SMILES string
Oc1ccc(C=O)cc1O
InChI
1S/C7H6O3/c8-4-5-1-2-6(9)7(10)3-5/h1-4,9-10H
InChI key
IBGBGRVKPALMCQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
comparable product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of ethnopharmacology, 136(1), 129-136 (2011-04-30)
To evaluate the pharmacokinetics of phenolic compounds after oral administration of Danshen extract in rat brain. Blood and brain microdialysis probes were inserted into jugular vein and cerebral cortex of rat under anesthesia and perfused with ringer's solution at the
Journal of ethnopharmacology, 133(2), 735-742 (2010-11-16)
We aimed to detect optimal ratio of cardioprotection-dependent absorbed bioactive compounds (ABCs) as quality control of guan-xin-er-hao (GXEH) formula extracted by various processings. Ferulic acid (F), tanshinol (T), hydroxysafflor yellow A (A), protocatechualdehyde (P) and paeoniflorin (E) in GXEH formula
Journal of nutritional science and vitaminology, 57(1), 118-122 (2011-04-23)
Although various biological activities of Phellinus gilvus (PG) have been reported, the active compounds responsible for these effects are not known. Here, we evaluated the activity of various solvent extracts of PG, and found the ethyl acetate extract (Fd) to
Basic & clinical pharmacology & toxicology, 110(4), 384-389 (2011-11-05)
Recent studies have demonstrated that nuclear factor-κB (NF-κB) and high-mobility group box 1 (HMGB1) are associated with the pathophysiology of sepsis. The present study was carried out to investigate the effects of protocatechuic aldehyde (PA) on an experimental model of
Acta pharmacologica Sinica, 31(5), 638-646 (2010-04-07)
To investigate the pharmacokinetic interactions induced by content variation of the main water-soluble components of Danshen injection in rats. Intravenous Danshen injection (control) or Danshen injection with danshensu (DSS), protocatechuic aldehyde (PAL), salvianolic acid A (Sal A) or salvianolic acid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service