Merck
  • Pharmacokinetic interactions induced by content variation of major water-soluble components of Danshen preparation in rats.
Pharmacokinetic interactions induced by content variation of major water-soluble components of Danshen preparation in rats.
Acta pharmacologica Sinica (2010-04-07)
Bo-bo Chang, Lin Zhang, Wan-wen Cao, Yuan Cao, Wen-liang Yang, Yan Wang, Yuan-cheng Chen, Xiao-quan Liu
ABSTRACT
To investigate the pharmacokinetic interactions induced by content variation of the main water-soluble components of Danshen injection in rats. Intravenous Danshen injection (control) or Danshen injection with danshensu (DSS), protocatechuic aldehyde (PAL), salvianolic acid A (Sal A) or salvianolic acid B (Sal B) were administered to female Sprague Dawley rats. Plasma concentrations of DSS, Sal A, PAL and its oxidative metabolite protocatechuic acid (PA) were analyzed simultaneously with LC-MS/MS; concentrations of Sal B were determined by the LC-MS method. Non-compartmental pharmacokinetic parameters were calculated and compared for identifying the pharmacokinetic interactions among these components. Compared with the control group, the DSS, Sal A, and Sal B groups had significant increases in AUC(0-infinity) in response to elevated concentrations of PAL (by 78.1%, 51.0%, and 82.9%, respectively), while the clearances (CL) were markedly reduced (by 42.5%, 32.9%, and 46.8%, respectively). Similarly, Sal A increased the AUC(0-infinity) of DSS and Sal B (26.7% and 82.4%, respectively) and substantially decreased their clearances (21.4% and 45.6%, respectively). In addition, the pharmacokinetics of DSS and Sal B were significantly affected by the content variation of the other major components; the AUC(0-infinity) increased by 45.1% and 52.1%, respectively, the CL dropped by 29.6% and 27.1%, respectively, and the T(1/2) was decreased by 22.0% and 19.6%, respectively. Complex, extensive pharmacokinetic interactions were observed among the major water-soluble constituents in the Danshen injection. The content variation of PAL had the most significant effect on the pharmacokinetic behaviors of other major constituents. Furthermore, the pharmacokinetics of DSS and Sal B were the most susceptible to the content change of other components.
Sigma-Aldrich
3,4-Dihydroxybenzaldehyde, purum, ≥97.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
3,4-Dihydroxybenzaldehyde, 97%
Pricing and availability is not currently available.