Skip to Content
Merck
All Photos(1)

Key Documents

419117

Sigma-Aldrich

Poly(maleic anhydride-alt-1-octadecene)

average Mn 30,000-50,000, powder (Free-flowing)

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002

form

powder (Free-flowing)

mol wt

average Mn 30,000-50,000

acid number

310‑315 mg KOH/g

transition temp

softening point 120-130 °C

solubility

H2O: insoluble
hydrocarbons: soluble

density

0.97 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

Poly(maleic anhydride-alt-1-octadecene) is a non-polar polymer.[1]

Application

Poly(maleic anhydride-alt-1-octadecene) (PMHC18) may be used to water-solubilize nanoparticles.[2] PMHC18 may be used to improve the pour point (PP) properties of palm oil methyl esters for its application as a vegetable oil-based biodiesel.[1] In vivo behaviors of single-walled nanotubes (SWNTs) can be fine tuned by coating SWNTs with polyethylene glycol (PEG) grafted PMHC18 polymers.[3]
Release coatings, anti-corrosive, thickening agent, chelant/surfactant and cross-linker for epoxy or alcohol functional polymers.

Features and Benefits

Readily derivatized to acid salt, amide, ester or imide. Alternating polymer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Water solubilization of hydrophobic nanocrystals by means of poly (maleic anhydride-alt-1-octadecene).
Di Corato R, et al.
Journal of Materials Chemistry, 18(17), 1991-1996 (2008)
Optimization of surface chemistry on single-walled carbon nanotubes for in vivo photothermal ablation of tumors.
Liu X, et al.
Biomaterials, 32(1), 144-151 (2011)
The effect of polymers and surfactants on the pour point of palm oil methyl esters.
Sern C H, et al.
European Journal of Lipid Science and Technology, 109(4), 440-444 (2007)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service